Paracetamol ≥98.0%

Supplier: TCI America

Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP

H0190-25G H0190-500G
TCH0190-025GEA 42.05 USD
TCH0190-025G TCH0190-500G
Paracetamol ≥98.0%
Paracetamol

CAS Number: 103-90-2 MDL Number: MFCD00002328 Molecular Formula: C8H9NO2 Molecular Weight: 151.17 Purity/Analysis Method: 98.0% (HPLC,N) Form: Crystal Melting point (°C): 169


Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Other Pharmaceutical Ingredients Other Product Names / Synonyms: 4-Acetamidophenol, Acetaminophen, Paracetamol

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 388
Melting Pt: 168
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 13,00048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=549e8b15a5028585d5868d656cef30642728f1ee&vid=549e8b15a5028585d5868d656cef30642728f1ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR