1-Hexene ≥97.0%

Supplier: TCI America
Danger

H0121-500ML H0121-25ML
TCH0121-500MLEA 32.21 USD
TCH0121-500ML TCH0121-25ML
1-Hexene ≥97.0%
1-Hexene

CAS Number: 592-41-6 MDL Number: MFCD00009505 Molecular Formula: C6H12 Molecular Weight: 84.16 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 64 Melting point (°C): -140 Flash Point (°C): -31 Specific Gravity (20/20): 0.67

Formula: C₆H₁₂
Boiling Pt: 64 °C
Melting Pt: –140 °C
Density: 0.67 (20 °C)
Flash Pt: –31 °C
Storage Temperature: Ambient
MDL Number: MFCD00009505
CAS Number: 592-41-6
EINECS: 209-753-1
UN: 2370
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5089733fc44ecc2421cd24c916ba9a9beaf75936&vid=357210d7fbd5035e487da9feafd3dd81c7a9b0f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR