Methyl hexanoate ≥98.0%

Supplier: TCI America

Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester

H0111-25ML H0111-500ML
TCH0111-025MLEA 25 USD
TCH0111-025ML TCH0111-500ML
Methyl hexanoate ≥98.0%
Methyl hexanoate

CAS Number: 106-70-7 MDL Number: MFCD00009510 Molecular Formula: C7H14O2 Molecular Weight: 130.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 150 Melting point (°C): -71 Flash Point (°C): 46 Specific Gravity (20/20): 0.89


Product Categories: Bioscience, Lipids Other Product Names / Synonyms: Hexanoic Acid Methyl Ester

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 151
Melting Pt: -63
Density: 0.89 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00009510
CAS Number: 106-70-7
EINECS: 203-425-1
UN: 3272
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dce5981f4ba4497ca8ac146744fb327656110783&vid=dce5981f4ba4497ca8ac146744fb327656110783&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR