1-Cetylpyridinium chloride monohydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate

H0078-500G H0078-25G H0078-100G
TCH0078-500GEA 117 USD
TCH0078-500G TCH0078-25G TCH0078-100G
1-Cetylpyridinium chloride monohydrate ≥98.0% (by HPLC, titration analysis)
1-Cetylpyridinium chloride monohydrate

CAS Number: 6004-24-6 MDL Number: MFCD00149977 Molecular Formula: C21H38ClN Molecular Weight: 339.99 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 84


Product Categories: Materials Chemistry, Cationic Surfactants Other Product Names / Synonyms: Cetylpyridinium Chloride

Formula: C₂₁H₃₈ClN·1H₂O
MW: 358.01 g/mol
Melting Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00149977
CAS Number: 6004-24-6
EINECS: 204-593-9
UN: 2811
ADR: 6.1,I
Merck Index: 13,02041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=431ffd811ccac842483ebf89eff5ad8b9f9da650&vid=431ffd811ccac842483ebf89eff5ad8b9f9da650&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR