Genistein ≥98.0% (by HPLC)
Supplier: TCI America
CAS Number: 446-72-0
MDL Number: MFCD00016952
Molecular Formula: C15H10O5
Molecular Weight: 270.24
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals), Epigenetics Research Reagents
Other Product Names / Synonyms: 4',5,7-Trihydroxyisoflavone
Formula:
C₁₅H₁₀O₅ Melting Pt: 297…298 °C |
MDL Number:
MFCD00016952 CAS Number: 446-72-0 EINECS: 207-174-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...