Dimethyl fumarate ≥98.0%

Supplier: TCI America

Synonyms: Fumaric acid dimethyl ester

F0069-25G F0069-500G
TCF0069-025GEA 35.82 USD
TCF0069-025G TCF0069-500G
Dimethyl fumarate ≥98.0%
Dimethyl fumarate

CAS Number: 624-49-7 MDL Number: MFCD00064438 Molecular Formula: C6H8O4 Molecular Weight: 144.13 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 205 Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction), Materials Chemistry, Copolymerization Monomers, Bioscience, Immunosuppressant Ingredients Other Product Names / Synonyms: Fumaric Acid Dimethyl Ester

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192
Melting Pt: 102
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064438
CAS Number: 624-49-7
EINECS: 210-849-0
Merck Index: 13,04308
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=347c694ca807cb29d6a611fb07b55460e096b661&vid=347c694ca807cb29d6a611fb07b55460e096b661&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR