Ethyl formate ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Formic acid ethyl ester
F0053-25ML
F0053-500ML
TCF0053-25MLEA
32.42
USD
TCF0053-25ML
TCF0053-500ML
Ethyl formate ≥98.0%
Ethyl formate
CAS Number: 109-94-4
MDL Number: MFCD00003294
Molecular Formula: C3H6O2
Molecular Weight: 74.08
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: Formic Acid Ethyl Ester
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 54 °C Melting Pt: –80 °C Density: 0.92 (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003294 CAS Number: 109-94-4 EINECS: 203-721-0 UN: 1190 ADR: 3,II Merck Index: 13,03841 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...