Flavone ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenylchromen-4-one

F0015-1G F0015-5G
TCF0015-001GEA 24 USD
TCF0015-001G TCF0015-005G
Flavone ≥98.0%
Flavone

CAS Number: 525-82-6 MDL Number: MFCD00006825 Molecular Formula: C15H10O2 Molecular Weight: 222.24 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 185 Melting point (°C): 95

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 185 °C
Melting Pt: 96
MDL Number: MFCD00006825
CAS Number: 525-82-6
EINECS: 208-383-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0c3c725008417ce95fb91c99a72597d6dc259b&vid=5f0c3c725008417ce95fb91c99a72597d6dc259b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR