Eriochrome Black T

Supplier: TCI America
Warning

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1)Eriochrome black T, IndicatorSodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonateEBTSolochrome Black TSodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

E0487-5G
TCE0487-005GEA 43.21 USD
TCE0487-005G
Eriochrome Black T
Eriochrome Black T

CAS Number: 1787-61-7
MDL Number: MFCD00003935
Molecular Formula: C20H13N3O7S
Molecular Weight: 461.38
Form: Crystal
Color: Deep Yellow Red


Other Product Names / Synonyms: Mordant Black 11

Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III
https://us-prod2.vwr.com/stibo/web/std.lang.all/80/01/28968001.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR