2-Ethylaniline ≥98.0%
Supplier: TCI America
|
Warning
|
E0060-500ML
E0060-25ML
TCE0060-500MLEA
147
USD
TCE0060-500ML
TCE0060-25ML
2-Ethylaniline ≥98.0%
2-Ethylaniline
CAS Number: 578-54-1
MDL Number: MFCD00007751
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method:
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 210 Melting Pt: -46 Density: 0.98 (20 °C) Flash Pt: 99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007751 CAS Number: 578-54-1 EINECS: 209-424-2 UN: 2273 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...