Propylene oxide ≥99.0%

Supplier: TCI America
Danger

Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide

E0016-500ML E0016-25ML
TCE0016-500MLEA 44.46 USD
TCE0016-500ML TCE0016-25ML
Propylene oxide ≥99.0%
Propylene oxide

CAS Number: 75-56-9 MDL Number: MFCD00005126 Molecular Formula: C3H6O Molecular Weight: 58.08 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 34 Flash Point (°C): -37 Specific Gravity (20/20): 0.83


Product Categories: Materials Chemistry, Monomers, Bioscience, Treatment Agents (Staining) Other Product Names / Synonyms: 1,2-Epoxypropane

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34.3
Melting Pt: -112
Density: 0.83 (20 °C)
Flash Pt: –37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005126
CAS Number: 75-56-9
EINECS: 200-879-2
UN: 1280
ADR: 3 I
Merck Index: 12,08041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=324f366e85dbe7852ab02813d82f45e928419d63&vid=324f366e85dbe7852ab02813d82f45e928419d63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR