6-Chloro-1,3-dimethyluracil ≥98.0%
Supplier: TCI America
Synonyms:
6-Chloro-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 1,3-Dimethyl-6-chlorouracil
D1619-5G
D1619-25G
TCD1619-5GEA
61.7
USD
TCD1619-5G
TCD1619-025G
6-Chloro-1,3-dimethyluracil ≥98.0%
6-Chloro-1,3-dimethyluracil
CAS Number: 6972-27-6
MDL Number: MFCD00038066
Molecular Formula: C6H7ClN2O2
Molecular Weight: 174.58
Purity/Analysis Method:
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Chloro-1,3-dimethyluracil
Formula:
C₆H₇ClN₂O₂ MW: 174.59 g/mol Melting Pt: 113 °C |
MDL Number:
MFCD00038066 CAS Number: 6972-27-6 EINECS: 230-205-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...