3,4-Dimethoxyaniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D1485-25G D1485-500G
TCD1485-025GEA 32.56 USD
TCD1485-025G TCD1485-500G
3,4-Dimethoxyaniline ≥98.0% (by GC, titration analysis)
3,4-Dimethoxyaniline

CAS Number: 6315-89-5 MDL Number: MFCD00008394 Molecular Formula: C8H11NO2 Molecular Weight: 153.18 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 180 Melting point (°C): 87 Flash Point (°C): 110


Other Product Names / Synonyms: 4-Aminoveratrole

Formula: C₈H₁₁NO₂
Boiling Pt: 180 °C
Melting Pt: 87 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3e96c2b8a20e041cf6a44653108d92b5074a3b1&vid=e4a94589cccce0b6306f499e50d93f8c197a80f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR