Dodecyltrimethylammonium chloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: (1-Dodecyl)trimethylammonium chloride, N,N,N-Trimethyldodecan-1-aminium chloride, DTAC

D1016-500G D1016-25G
TCD1016-500GEA 450 USD
TCD1016-500G TCD1016-25G
Dodecyltrimethylammonium chloride ≥97.0% (by titrimetric analysis)
Dodecyltrimethylammonium chloride

CAS Number: 112-00-5 MDL Number: MFCD00041974 Molecular Formula: C15H34ClN Molecular Weight: 263.89 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 246 Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry Other Product Names / Synonyms: Lauryltrimethylammonium Chloride, DTAC

Formula: C₁₅H₃₄ClN
MW: 263.89 g/mol
Melting Pt: 246 °C
MDL Number: MFCD00041974
CAS Number: 112-00-5
EINECS: 203-927-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ea00cf360ab1ad0465e804b637abe07e6201e53&vid=2ea00cf360ab1ad0465e804b637abe07e6201e53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR