Barium diphenylamine-4-sulfonate ≥98.0%

Supplier: TCI America
Warning

Synonyms: N-Phenylsulfanilic acid barium salt, Diphenylamine-4-sulfonic acid barium salt, Diphenylamine-4-sulphonic acid barium salt, N-Phenylsulphanilic acid barium salt

D0876-1G D0876-25G D0876-5G
TCD0876-001GEA 24.42 USD
TCD0876-001G TCD0876-025G TCD0876-5G
Barium diphenylamine-4-sulfonate ≥98.0%
Barium diphenylamine-4-sulphonate

CAS Number: 6211-24-1 MDL Number: MFCD00007497 Molecular Formula: C12H11NO3S Molecular Weight: 633.88 Purity/Analysis Method: 98.0% (N) Form: Crystal


Other Product Names / Synonyms: Diphenylamine-4-sulfonic Acid Barium Salt, N-Phenylsulfanilic Acid Barium Salt

Formula: C₂₄H₂₀BaN₂O₆S₂
MW: 633.89 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007497
CAS Number: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ae72a758370870c065973fbd83c1f929499c89a&vid=3ae72a758370870c065973fbd83c1f929499c89a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR