Dimethyl sulfoxide 99,0

Supplier: TCI America

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

D0798-500G D0798-25G
TCD0798-500GEA 40.26 USD
TCD0798-500G TCD0798-025G
Dimethyl sulfoxide 99,0
Dimethyl sulphoxide

CAS Number: 67-68-5
MDL Number: MFCD00002089
Molecular Formula: C2H6OS
Molecular Weight: 78.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 95
Freezing point (°C): 18
Specific Gravity (20/20): 1.10


Product Categories: Synthetic Organic Chemistry, Sulfoxides, C-H Bond Activation Reaction, Bioscience, Treatment Agents (Staining), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: DMSO

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.10 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0688decad5c8afa4e8a1177979cd07eb3c07316&vid=b69a1fae7562bd9674ce37578ffda941dd0f0b16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR