3,4-Xylenol ≥98.0%

Supplier: TCI America
Danger

D0777-500G D0777-25G
TCD0777-500GEA 181 USD
TCD0777-500G TCD0777-25G
3,4-Xylenol ≥98.0%
3,4-Xylenol

CAS Number: 95-65-8 MDL Number: MFCD00002304 Molecular Formula: C8H10O Molecular Weight: 122.17 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 225 Melting point (°C): 66 Flash Point (°C): 110


Other Product Names / Synonyms: o-Xylenol

Formula: C₈H₁₀O
Boiling Pt: 225 °C
Melting Pt: 66 °C
Density: 1.023 g/cm³ (17 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002304
CAS Number: 95-65-8
EINECS: 202-439-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8eab18968e017905e8cf51e3e1d64289d6e65bb&vid=309cd43441e4af11b2a904a449e8602c8801673e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR