2,3-Dimethyl-2-butene ≥96.0%
Supplier: TCI America
Synonyms:
Tetramethylethylene
D0693-25ML
D0693-500ML
TCD0693-025MLEA
79
USD
TCD0693-025ML
TCD0693-500ML
2,3-Dimethyl-2-butene ≥96.0%
2,3-Dimethyl-2-butene
CAS Number: 563-79-1
MDL Number: MFCD00008897
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method:
Other Product Names / Synonyms: Tetramethylethylene
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 73 Melting Pt: -75 Density: 0.71 (20 °C) Flash Pt: –16 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008897 CAS Number: 563-79-1 EINECS: 209-263-8 UN: 3295 ADR: 3 II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...