N,N-Dibenzylaniline ≥99.0%

Supplier: TCI America

D0154-25G D0154-500G
TCD0154-025GEA 33.5 USD
TCD0154-025G TCD0154-500G
N,N-Dibenzylaniline ≥99.0%
N,N-Dibenzylaniline

CAS Number: 91-73-6 MDL Number: MFCD00022015 Molecular Formula: C20H19N Molecular Weight: 273.38 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 180 Melting point (°C): 70


Other Product Names / Synonyms: N-Phenyldibenzylamine

Formula: (C₆H₅)N(CH₂C₆H₅)₂
Boiling Pt: 180 °C
Melting Pt: 70 °C
Density: 1.1 g/cm³ (20 °C)
MDL Number: MFCD00022015
CAS Number: 91-73-6
EINECS: 202-093-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=654b63eff212345afd69773bf5cb5c1b0aaa2577&vid=dbfb6f7b8ea302d6f26a345cfe8a55c9b5e51626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR