5-Chlorovaleronitrile ≥96.0%

Supplier: TCI America
Warning

Synonyms: 5-Chloropentanenitrile

C1421-250G C1421-25G
TCC1421-250GEA 222 USD
TCC1421-250G TCC1421-25G
5-Chlorovaleronitrile ≥96.0%
5-Chlorovaleronitrile

CAS Number: 6280-87-1 MDL Number: MFCD00001977 Molecular Formula: C5H8ClN Molecular Weight: 117.58 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling point (°C): 118 Flash Point (°C): 98 Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 4-Chlorobutyl Cyanide, 5-Chloropentanenitrile

Formula: C₅H₈ClN
Boiling Pt: 118 °C
Density: 1.05 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00001977
CAS Number: 6280-87-1
EINECS: 228-481-4
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f6f34f6704cc60c620347b5bb871ef8385d8c7&vid=f0f6f34f6704cc60c620347b5bb871ef8385d8c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR