2-Chloroethyl methyl ether ≥98.0%

Supplier: TCI America

C1173-25ML C1173-500ML C1173-100ML
TCC1173-025MLEA 36 USD
TCC1173-025ML TCC1173-500ML TCC1173-100ML
2-Chloroethyl methyl ether ≥98.0%
2-Chloroethyl methyl ether

CAS Number: 627-42-9 MDL Number: MFCD00000972 Molecular Formula: C3H7ClO Molecular Weight: 94.54 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 92 Flash Point (°C): 8 Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-Methoxyethyl Chloride

Formula: C₃H₇ClO
Boiling Pt: 92 °C
Melting Pt: –55…55 °C
Density: 1.04 (20 °C)
Flash Pt: 8 °C
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cd3ed0b93544bf2fddc8baaff46c9d791934be7&vid=9cd3ed0b93544bf2fddc8baaff46c9d791934be7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR