1,4-Cyclohexadiene ≥98.0% stabilised

Supplier: TCI America

Synonyms: Cyclohexa-1,4-diene

C0468-10ML C0468-25ML
TCC0468-010MLEA 96.67 USD
TCC0468-010ML TCC0468-25ML
1,4-Cyclohexadiene ≥98.0% stabilised
1,4-Cyclohexadiene

(stabilized with BHT) CAS Number: 628-41-1 MDL Number: MFCD00001535 Molecular Formula: C6H8 Molecular Weight: 80.13 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 88 Melting point (°C): -49 Flash Point (°C): -11 Specific Gravity (20/20): 0.86 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ligands Other Product Names / Synonyms: 1,4-Dihydrobenzene

Formula: C₆H₈
MW: 80.13 g/mol
Boiling Pt: 82
Melting Pt: -49.2
Density: 0.86 (20 °C)
Flash Pt: –11 °C
MDL Number: MFCD00001535
CAS Number: 628-41-1
EINECS: 211-043-1
UN: 3295
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ed860379852b0debcbf1d4dc93111abaf7a719d&vid=8ed860379852b0debcbf1d4dc93111abaf7a719d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR