Coumarin ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1-Benzopyran-2-one, 2H-Chromen-2-one, 1,2-Benzopyrone

C0395-500G C0395-25G
TCC0395-500GEA 87 USD
TCC0395-500G TCC0395-25G
Coumarin ≥99.0%
Coumarin

CAS Number: 91-64-5 MDL Number: MFCD00006850 Molecular Formula: C9H6O2 Molecular Weight: 146.15 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 298 Melting point (°C): 70 Flash Point (°C): 151 Lambda max.: 273 nm


Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals) Other Product Names / Synonyms: Cumarin

Formula: C₉H₆O₂
MW: 146.15 g/mol
Boiling Pt: 298 °C
Melting Pt: 70 °C
Density: 0.935 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00006850
CAS Number: 91-64-5
EINECS: 202-086-7
UN: 2811
ADR: 6.1,III
Merck Index: 13,02588
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4f2b4b1d8c967c55d5bdc79bf2abfbefe897711&vid=a4f2b4b1d8c967c55d5bdc79bf2abfbefe897711&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR