3-Chloro-α,α,α-trifluorotoluene ≥90.0%
Supplier: TCI America
Synonyms:
1-Chloro-3-(trifluoromethyl)benzene, 3-Chlorobenzotrifluoride
C0304-500G
C0304-25G
TCC0304-500GEA
330
USD
TCC0304-500G
TCC0304-25G
3-Chloro-α,α,α-trifluorotoluene ≥90.0%
3-Chloro-α,α,α-trifluorotoluene
CAS Number: 98-15-7
MDL Number: MFCD00000594
Molecular Formula: C7H4ClF3
Molecular Weight: 180.55
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Chloro-3-(trifluoromethyl)benzene, 3-Chloro-alpha,alpha,alpha-trifluorotoluene
Formula:
C₇H₄ClF₃ Boiling Pt: 139 °C Melting Pt: –56 °C Density: 1.35 (20 °C) Flash Pt: 36 °C |
MDL Number:
MFCD00000594 CAS Number: 98-15-7 EINECS: 202-642-9 UN: 2234 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...