4-Chloro-3,5-xylenol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol

C0161-25G C0161-500G
TCC0161-025GEA 25 USD
TCC0161-025G TCC0161-500G
4-Chloro-3,5-xylenol ≥98.0%
4-Chloro-3,5-xylenol

CAS Number: 88-04-0 MDL Number: MFCD00002324 Molecular Formula: C8H9ClO Molecular Weight: 156.61 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 246 Melting point (°C): 115 Flash Point (°C): 138


Other Product Names / Synonyms: 4-Chloro-3,5-xylenol, PCMX

Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246
Melting Pt: 115 °C
Density: 1.364 g/cm³ (20 °C)
Flash Pt: 138 °C
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
Merck Index: 13,02194
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4153c9be3d6a0c27f29657b0665ab6fdc4140fe2&vid=4153c9be3d6a0c27f29657b0665ab6fdc4140fe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR