Bromoacetonitrile ≥97.0%

Supplier: TCI America
Danger

B1058-500G B1058-25G B1058-100G
TCB1058-500GEA 760 USD
TCB1058-500G TCB1058-25G TCB1058-100G
Bromoacetonitrile ≥97.0%
Bromoacetonitrile

CAS Number: 590-17-0 MDL Number: MFCD00001884 Molecular Formula: C2H2BrN Molecular Weight: 119.95 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 150 Flash Point (°C): 110 Specific Gravity (20/20): 1.80

Formula: BrCNCH₂
MW: 119.95 g/mol
Boiling Pt: 150
Melting Pt: -36
Density: 1.80 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001884
CAS Number: 590-17-0
EINECS: 209-672-1
UN: 3276
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a886a303fdec4f98bb9a18ac3a37fe08e5533052&vid=a886a303fdec4f98bb9a18ac3a37fe08e5533052&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR