4-tert-Butylpyrocatechol ≥98.0%

Supplier: TCI America
Danger

Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

B0739-500G B0739-25G
TCB0739-500GEA 85.47 USD
TCB0739-500G TCB0739-25G
4-tert-Butylpyrocatechol ≥98.0%
4-tert-Butylpyrocatechol

CAS Number: 98-29-3 MDL Number: MFCD00002201 Molecular Formula: C10H14O2 Molecular Weight: 166.22 Purity/Analysis Method: 98.0% (GC) Form: Crystal Boiling point (°C): 285 Melting point (°C): 58 Flash Point (°C): 130


Product Categories: Materials Chemistry, Polymerization Inhibitors Other Product Names / Synonyms: 4-tert-Butylcatechol, TBC

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 285 °C
Melting Pt: 58 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e17cca7b3366c7ec4a08328cc82bc55f5384626a&vid=e17cca7b3366c7ec4a08328cc82bc55f5384626a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR