N-Bromosuccinimide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 128-08-5
MDL Number: MFCD00005510
Molecular Formula: C4H4BrNO2
Molecular Weight: 177.99
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Oxidation, Halogenation, Glycosylation, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: NBS
Formula:
C₄H₄BrNO₂ Boiling Pt: 339 °C (1013 hPa) Melting Pt: 176 °C Density: 2.098 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005510 CAS Number: 128-08-5 EINECS: 204-877-2 UN: 3261 ADR: 8,III Merck Index: 13,01423 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...