Anthrone ≥98.0%

Supplier: TCI America
Warning

Synonyms: Anthracen-9(10H)-one

A0509-25G A0509-5G
TCA0509-025GEA 340.91 USD
TCA0509-025G TCA0509-5G
Anthrone ≥98.0%
Anthrone

CAS Number: 90-44-8 MDL Number: MFCD00001187 Molecular Formula: C14H10O Molecular Weight: 194.23 Purity/Analysis Method: 98.0% (GC) Form: Crystal Melting point (°C): 156

Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Melting Pt: 156 °C
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=382326923f8de38fc75922f5919e9b9c43f2d069&vid=382326923f8de38fc75922f5919e9b9c43f2d069&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR