4-Aminosalicylic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Amino-2-hydroxybenzoic acid

A0420-25G A0420-100G
TCA0420-025GEA 146.01 USD
TCA0420-025G TCA0420-100G
4-Aminosalicylic acid ≥98.0%
4-Aminosalicylic acid

CAS Number: 65-49-6 MDL Number: MFCD00007789 Molecular Formula: C7H7NO3 Molecular Weight: 153.14 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 151


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents Other Product Names / Synonyms: 4-Amino-2-hydroxybenzoic Acid

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 150
Density: 1.545 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007789
CAS Number: 65-49-6
EINECS: 200-613-5
Merck Index: 13,00476
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17494fb417100dc2c1dad1ea34b3ce499c6c96f8&vid=17494fb417100dc2c1dad1ea34b3ce499c6c96f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR