5-Aminosalicylic acid ≥98.0%
Supplier: TCI America
Synonyms:
Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
CAS Number: 89-57-6
MDL Number: MFCD00007877
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method:
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immune System, Inflammation, Allergy Research Reagents, Immunosuppressant Ingredients, Anti-rheumatoid Arthritis Ingredients, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 5-Amino-2-hydroxybenzoic Acid, Mesalamine, Mesalazine
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Boiling Pt: 347 Melting Pt: 280 Density: 1.568 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007877 CAS Number: 89-57-6 EINECS: 201-919-1 Merck Index: 12,05964 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...