Cysteaminium chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Cysteamine Hydrochloride
, Mercaptoethylamine-HCl
, β-Mercaptoethylamine hydrochloride
, H-Cys-NH2·HCl
, 2-Aminoethanethiol hydrochloride
, 2-MEA
, 2-Mercaptoethylamine hydrochloride
CAS Number: 156-57-0
MDL Number: MFCD00012904
Molecular Formula: C2H7NS
Molecular Weight: 113.60
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 68
Flash Point (°C): 66
Storage Temperature: 0-10°C
Product Categories: Bioscience, Reagents for Inflammation & Immunity Disorder Models, Antioxidants, Functionality Creating Reagents
Other Product Names / Synonyms: Cysteamine Hydrochloride, 2-Mercaptoethylamine Hydrochloride
Formula:
C₂H₈ClNS MW: 113.61 g/mol Melting Pt: 68 °C Flash Pt: 66 °C |
MDL Number:
MFCD00012904 CAS Number: 156-57-0 EINECS: 205-858-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...