2-Amino-4-chlorophenol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Chloro-2-hydroxyaniline

A0284-500G A0284-25G
TCA0284-500GEA 202 USD
TCA0284-500G TCA0284-25G
2-Amino-4-chlorophenol ≥98.0%
2-Amino-4-chlorophenol

CAS Number: 95-85-2 MDL Number: MFCD00007694 Molecular Formula: C6H6ClNO Molecular Weight: 143.57 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 139 Flash Point (°C): 170

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 139 °C
Flash Pt: 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00007694
CAS Number: 95-85-2
EINECS: 202-458-9
UN: 2673
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6662ebba3635f1c0238e0fe5a3a9ee853d6e04f3&vid=6662ebba3635f1c0238e0fe5a3a9ee853d6e04f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR