2',3'-Dideoxycytidine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine
L10619-MD
L10619-MF
AAAL10619-MDEA
245
USD
AAAL10619-MD
AAAL10619-MF
2',3'-Dideoxycytidine 98+%
2',3'-Dideoxycytidine
Purity: 98+%
CAS No.: 7481-89-2
Molecular Formula: C9H13N3O3
Formula Weight: 211.22
Storage Temperature: 2°C to 8°C
Melting Point: 216-220°
Beilstein Reference: 654956
Merck Reference: 14,10109
Optical Rotation: +90° (c=0.5 in water)
MDL No.: MFCD00012188
Formula:
C₉H₁₃N₃O₃ MW: 211.22 g/mol Melting Pt: 216…220 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012188 CAS Number: 7481-89-2 Merck Index: 14,10109 |
Specification Test Results
Optical rotation | +90° (c=0.5 in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...