3-Bromo-4-fluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B25610-06
B25610-14
AAB25610-03EA
37.62
USD
AAB25610-03
AAB25610-06
AAB25610-14
3-Bromo-4-fluoroaniline 98%
3-Bromo-4-fluoroaniline
Purity: 98%
CAS No.: 656-64-4
Molecular Formula: C6H5BrFN
Formula Weight: 190.01
Melting Point: 34-36°
Boiling Point: 235°
UN No.: 2811
MDL No.: MFCD03788559
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 235 °C Melting Pt: 34…36 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788559 CAS Number: 656-64-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...