Ferrocenecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ferrocene monocarboxylic acid

A16874-06 A16874-03
AAAA16874-06EA 164.74 USD
AAAA16874-06 AAAA16874-03 AAAA16874-14
Ferrocenecarboxylic acid 98%
Ferrocenecarboxylic acid

Purity: 98% CAS No.: 1271-42-7 Molecular Formula: C11H10FeO2 Formula Weight: 230.05 Melting Point: ca 210° dec. MDL No.: MFCD00001430

Formula: C₁₁H₁₀FeO₂
Melting Pt: ca 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001430
CAS Number: 1271-42-7
EINECS: 215-040-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR