Lead ≥99.95% (metals basis), Powder -100 mesh

Supplier: Thermo Scientific Chemicals
Danger

045602-30 045602-A1
AA45602-30EA 48.11 USD
AA45602-30 AA45602-A1
Lead ≥99.95% (metals basis), Powder -100 mesh
Lead

Purity: 99.95% CAS No.: 7439-92-1 Molecular Formula: Pb Physical Form: Atomized powder Odor: Odorless UN No.: 3077 MDL No.: MFCD00134050

Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1749 °C (1013 hPa)
Melting Pt: 327,5 °C
Density: 11,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR