Copper ≥99% (metals basis), soluble -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
045073-30
045073-A1
AA45073-30EA
124
USD
AA45073-30
AA45073-A1
Copper ≥99% (metals basis), soluble -325 mesh
Copper
Purity: 99%
CAS No.: 7440-50-8
Molecular Formula: Cu
Formula Weight: 63.55
Physical Form: Flake, average thickness 1-2 microns
Density: 8.94
Melting Point: 1083°
Boiling Point: 2562°
UN No.: 3089
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Density: 8.94 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 UN: 3089 ADR: 4.1 III Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...