Ruthenium ≥99.99% (metals basis excluding Ca), powder, Premion® -200 mesh (max. 0.01% Ca)

Supplier: Thermo Scientific Chemicals
Warning

044879-03 044879-06 044879-14
AA44879-03EA 165 USD
AA44879-03 AA44879-06 AA44879-14
Ruthenium ≥99.99% (metals basis excluding Ca), powder, Premion® -200 mesh (max. 0.01% Ca)
Ruthenium

Purity: 99.99+% CAS No.: 7440-18-8 Appearance: Grey Odor: Odorless UN No.: 3089 MDL No.: MFCD00011207

Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR