N-Benzylacrylamide 96%

Supplier: Thermo Scientific Chemicals
Warning

L11450-03 L11450-06
AAAL11450-03EA 54.98 USD
AAAL11450-03 AAAL11450-06
N-Benzylacrylamide 96%
N-Benzylacrylamide

Purity: 96% CAS No.: 13304-62-6 Molecular Formula: CH2=CHCONHCH2C6H5 Formula Weight: 161.21 Melting Point: 59-62° Beilstein Reference: 2690053 MDL No.: MFCD00015333

Formula: CH₂=CHCONHCH₂C₆H₅
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00015333
CAS Number: 13304-62-6
EINECS: 603-701-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR