(±)-Naringenin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone, Naringenin

L09834-04 L09834-09
AAAL09834-04EA 46.47 USD
AAAL09834-04 AAAL09834-09
(±)-Naringenin 97%
(±)-Naringenin

Purity: 97% CAS No.: 67604-48-2 Molecular Formula: C15H12O5 Formula Weight: 272.26 Melting Point: 248-251° Merck Reference: 14,6424 MDL No.: MFCD00006844

Formula: C₁₅H₁₂O₅
MW: 272.25 g/mol
Melting Pt: 248…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00006844
CAS Number: 67604-48-2
EINECS: 266-769-1
Merck Index: 14,6424

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR