Hydrogen chloride 1 M in acetic acid
Supplier: Thermo Scientific Chemicals
|
Danger
|
H27347-AE
H27347-AU
AAH27347-AEEA
29.7
USD
AAH27347-AE
AAH27347-AU
Hydrogen chloride 1 M in acetic acid
Hydrogen chloride
CAS No.: 7647-01-0
Molecular Formula: HCl
Formula Weight: 36.46
Flash Point: 40°(104°F)
Odor: Acrid
UN No.: 2920
MDL No.: MFCD00011324
Formula:
HCl MW: 36.46 g/mol Density: 1.050 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...