p-Sexiphenyl 95%

Supplier: Thermo Scientific Chemicals

H26586-03 H26586-MD
AAH26586-03EA 516 USD
AAH26586-03 AAH26586-MD
p-Sexiphenyl 95%
p-Sexiphenyl

Purity: 95% CAS No.: 4499-83-6 Molecular Formula: C36H26 Formula Weight: 458.59 Melting Point: 400° MDL No.: MFCD00039559

Formula: C₃₆H₂₆
Melting Pt: >400 °C
Storage Temperature: Ambient
MDL Number: MFCD00039559
CAS Number: 4499-83-6
EINECS: 224-799-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR