6-Thioguanine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B21280-03
B21280-06
AAB21280-03EA
85.4
USD
AAB21280-03
AAB21280-06
6-Thioguanine 98%
6-Thioguanine
Purity: 98%
CAS No.: 154-42-7
Molecular Formula: C5H5N5S
Formula Weight: 167.19
Odor: Odorless
Melting Point:
Formula:
C₅H₅N₅S Melting Pt: >360 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233553 CAS Number: 154-42-7 EINECS: 205-827-2 UN: 2811 ADR: "6.1",III Merck Index: 14,9337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...