Ethyldiphenylphosphine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A18292-09
A18292-18
AAA18292-09EA
110
USD
AAA18292-09
AAA18292-18
Ethyldiphenylphosphine 98%
Ethyldiphenylphosphine
Purity: 98%
CAS No.: 607-01-2
Molecular Formula: CH3CH2P(C6H5)2
Formula Weight: 214.25
Odor: Stench
Boiling Point: 130-132°/1mm
UN No.: 3278
MDL No.: MFCD00015170
Formula:
C₁₄H₁₅P Boiling Pt: 130…132 °C/1 mm Density: 1.048 Storage Temperature: Ambient |
MDL Number:
MFCD00015170 CAS Number: 607-01-2 UN: 3334 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...