Calcium pantothenate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate
A16609-22
A16609-36
AAA16609-22EA
80
USD
AAA16609-22
AAAA16609-36
Calcium pantothenate 98%
Calcium pantothenate
Vitamin B-5
Purity: 98%
CAS No.: 331748-07-3
Molecular Formula: C18H32CaN2O10.xH2O
Formula Weight: 476.54(anhy)
Melting Point: ca 138° dec.
Beilstein Reference: 3769272
Optical Rotation: +27° (c=5 in water)
MDL No.: MFCD00002766
Formula:
C₁₈H₃₂CaN₂O₁₀ MW: 476.54 g/mol Melting Pt: ∼ 138 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002766 CAS Number: 137-08-6 EINECS: 205-278-9 Merck Index: 14,7015 |
Specification Test Results
Optical rotation | +27° (c=5 in water) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...