Ethoxyacetylene 50% (w/w) in hexane
Supplier: Thermo Scientific Chemicals
|
Danger
|
Fieser: 1,357 2,190 5,290
CAS No.: 927-80-0
Molecular Formula: C4H6O
Formula Weight: 70.09
Storage Temperature: 2°C to 8°C
Flash Point: -28°(-18°F)
Refractive Index: 1.3800
Odor: Recognizable
Boiling Point: 55-56°
Beilstein Reference: 741882
UN No.: 1993
MDL No.: MFCD00009247
Formula:
C₄H₆O Boiling Pt: 55…56 °C Density: 0.718 Flash Pt: -28 °C(-18 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009247 CAS Number: 927-80-0 EINECS: 213-164-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...