Quinidine

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (+)-Quinidine

A12559-06 A12559-14 A12559-03
AAA12559-06EA 65.4 USD
AAA12559-06 AAA12559-14 AAA12559-03
Quinidine
Quinidine

Antiarrhythmic agent that blocks the inward sodium channel


CAS No.: 56-54-2 Molecular Formula: C20H24N2O2 Formula Weight: 324.42 Melting Point: 171-173° Beilstein Reference: 91866 Optical Rotation: +260° (c=1 in ethanol) UN No.: 2811 MDL No.: MFCD00135581


Applications: Antiarrhythmic agent that blocks the inward sodium channel

Formula: C20H24N2O2
MW: 324.42 g/mol
Melting Pt: 171…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00135581
CAS Number: 56-54-2
EINECS: 200-279-0
UN: 1544
ADR: "6.1",III
Merck Index: 14,8060

Order Now

Specification Test Results

Optical rotation +260° (c=1 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR