Silicone oil high temperature , colourless liquid for oil baths

Supplier: Thermo Scientific Chemicals

Synonyms: Polyphenyl-methylsiloxane

045896-22 045896-36 045896-A4
AA45896-22EA 104.72 USD
AA45896-22 AA45896-36 AA45896-A4
Silicone oil high temperature , colourless liquid for oil baths
Silicone oil

Usable temperature range: 25 to 250 °C (open system) and 25 to 315 °C (closed system)


Refractive Index: 1.505


Applications: High temperature open system, closed system, circulating, closed loop, and heat transfer baths. High temperature fluids for laboratory research apparatus and instruments.

Density: 1.07 g/mL
Flash Pt: >300 °C(572 °F)
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Merck Index: 14,8495

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR