Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness 0.025 mm (0.001 in) Type 304
Supplier: Thermo Scientific Chemicals
|
Danger
|
041580-FF
041580-FI
041580-GH
AA41580-FFEA
34.5
USD
AA41580-FF
AA41580-FI
AA41580-GH
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), foil, Thickness 0.025 mm (0.001 in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...